Assay Details
| Gene Symbol: Cln5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033242.1 | NP_001028414.1 | 2 - 3 | 418 | 76 | |
| GenBank mRNA | AK032293.1 | - | 2 - 3 | 233 | 76 | |
| AK043247.1 | - | 2 - 3 | 342 | 76 | ||
| AK085741.1 | - | 2 - 3 | 419 | 76 | ||
| AK144635.1 | - | 2 - 3 | 309 | 76 | ||
| AK207183.1 | - | 1 - 2 | 70 | 76 | ||
| AK209323.1 | - | 1 - 2 | 70 | 76 | ||
| AK211228.1 | - | 1 - 2 | 70 | 76 | ||
| AK214368.1 | - | 1 - 2 | 70 | 76 | ||
| AK216934.1 | - | 1 - 2 | 70 | 76 | ||
| AK218788.1 | - | 1 - 2 | 70 | 76 | ||
| BC025487.1 | - | 2 - 3 | 275 | 76 | ||
| BC141314.1 | - | 2 - 3 | 312 | 9055926 | 76 | |
| BC141315.1 | - | 2 - 3 | 312 | 9055927 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cln5
Genomic Map