Assay Details
| Gene Symbol: Irg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008392.1 | NP_032418.1 | 1 - 2 | 66 | 75 | |
| GenBank mRNA | AK036446.1 | - | 1 - 2 | 102 | 75 | |
| AK089891.1 | - | 1 - 2 | 48 | 75 | ||
| AK152177.1 | - | 1 - 2 | 65 | 75 | ||
| AK152248.1 | - | 1 - 2 | 65 | 75 | ||
| AK152332.1 | - | 1 - 2 | 65 | 75 | ||
| AK152635.1 | - | 1 - 2 | 65 | 75 | ||
| AK153013.1 | - | 1 - 2 | 65 | 75 | ||
| AK172001.1 | - | 1 - 2 | 65 | 75 | ||
| AK172298.1 | - | 1 - 2 | 65 | 75 | ||
| L38281.1 | - | 1 - 2 | 283 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK036446.1, AK152177.1, AK152248.1, AK152332.1, AK152635.1, AK172001.1, AK172298.1, NM_008392.1 transcripts.
Set Membership: |
|

Irg1
Genomic Map