Assay Details
| Gene Symbol: Irg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008392.1 | NP_032418.1 | 3 - 4 | 321 | 68 | |
| GenBank mRNA | AK036446.1 | - | 3 - 4 | 357 | 68 | |
| AK089891.1 | - | 3 - 4 | 303 | 68 | ||
| AK152177.1 | - | 3 - 4 | 320 | 68 | ||
| AK152248.1 | - | 3 - 4 | 320 | 68 | ||
| AK152332.1 | - | 3 - 4 | 320 | 68 | ||
| AK152635.1 | - | 3 - 4 | 320 | 68 | ||
| AK153013.1 | - | 3 - 4 | 320 | 68 | ||
| AK172001.1 | - | 3 - 4 | 320 | 68 | ||
| AK172298.1 | - | 3 - 4 | 320 | 68 | ||
| L38281.1 | - | 3 - 4 | 538 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Irg1
Genomic Map