Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400020343.mm.1 |
NM_145465.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020343.mm.1 |
NM_145465.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020343.mm.1 |
TC1400002735.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400020340.mm.1 |
NM_145465.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020340.mm.1 |
NM_145465.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020340.mm.1 |
TC1400002735.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400020341.mm.1 |
NM_145465.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020341.mm.1 |
NM_145465.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020341.mm.1 |
TC1400002735.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400020344.mm.1 |
NM_145465.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020344.mm.1 |
NM_145465.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020344.mm.1 |
TC1400002735.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400020345.mm.1 |
NM_145465.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020345.mm.1 |
NM_145465.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020345.mm.1 |
TC1400002735.mm.1 |
MTA TC |