Assay Details
| Gene Symbol: Ubac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026861.2 | NP_081137.2 | 1 - 2 | 173 | 169 | |
| GenBank mRNA | BC024467.1 | - | 1 - 2 | 159 | 4975777 | 169 |
| BY080555.1 | - | 1 - 2 | 173 | 169 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC024467.1, NM_026861.2 transcripts.
Set Membership: |
|

Ubac2
Genomic Map