Assay Details
| Gene Symbol: Ubac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026861.2 | NP_081137.2 | 2 - 3 | 295 | 90 | |
| GenBank mRNA | AK049109.1 | - | 2 - 3 | 244 | 90 | |
| AK158177.1 | - | 2 - 3 | 244 | 90 | ||
| BC024467.1 | - | 2 - 3 | 281 | 4975777 | 90 | |
| BY080555.1 | - | 2 - 3 | 295 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ubac2
Genomic Map