Assay Details
| Gene Symbol: Ubac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026861.2 | NP_081137.2 | 4 - 5 | 526 | 70 | |
| GenBank mRNA | AK035384.1 | - | 1 - 2 | 101 | 70 | |
| AK049109.1 | - | 4 - 5 | 475 | 70 | ||
| AK158177.1 | - | 4 - 5 | 475 | 70 | ||
| BC024467.1 | - | 4 - 5 | 512 | 4975777 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubac2
Genomic Map