Assay Details
| Gene Symbol: Ubac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026861.2 | NP_081137.2 | 7 - 8 | 942 | 61 | |
| GenBank mRNA | AK035384.1 | - | 4 - 5 | 517 | 61 | |
| AK049109.1 | - | 7 - 8 | 891 | 61 | ||
| AK158177.1 | - | 7 - 8 | 891 | 61 | ||
| AK159755.1 | - | 2 - 3 | 300 | 61 | ||
| BC017635.1 | - | 1 - 2 | 104 | 61 | ||
| BC024467.1 | - | 7 - 8 | 928 | 4975777 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubac2
Genomic Map