Assay Details
| Gene Symbol: Sema3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028882.4 | NP_083158.3 | 9 - 10 | 1525 | 127 | |
| GenBank mRNA | AK028900.1 | - | 9 - 10 | 1307 | 127 | |
| AK031704.1 | - | 9 - 10 | 1137 | 127 | ||
| AK052671.1 | - | 9 - 10 | 1526 | 127 | ||
| AK153839.1 | - | 9 - 10 | 1186 | 127 | ||
| BC138131.1 | - | 9 - 10 | 1068 | 8861150 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sema3d
Genomic Map