Assay Details
| Gene Symbol: Heph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159627.1 | NP_001153099.1 | 6 - 7 | 1301 | 84 | |
| NM_001159628.1 | NP_001153100.1 | 6 - 7 | 1309 | 84 | ||
| NM_010417.2 | NP_034547.2 | 6 - 7 | 1309 | 84 | ||
| NM_181273.4 | NP_851790.1 | 6 - 7 | 1301 | 84 | ||
| GenBank mRNA | AF082567.1 | - | 6 - 7 | 1421 | 84 | |
| AK032895.1 | - | 6 - 7 | 1221 | 84 | ||
| AK081330.1 | - | 6 - 7 | 1277 | 84 | ||
| AK086923.1 | - | 6 - 7 | 1220 | 84 | ||
| AK129194.1 | - | 6 - 7 | 1257 | 84 | ||
| BC048237.1 | - | 6 - 7 | 1149 | 6474714 | 84 | |
| BC054442.1 | - | 6 - 7 | 1149 | 6474713 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Heph
Genomic Map