Assay Details
| Gene Symbol: Timp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011595.2 | NP_035725.1 | 3 - 4 | 761 | 76 | |
| GenBank mRNA | AB041611.2 | - | 3 - 4 | 618 | 76 | |
| AK083905.1 | - | 3 - 4 | 619 | 76 | ||
| AK128943.1 | - | 3 - 4 | 620 | 76 | ||
| AK147847.1 | - | 3 - 4 | 619 | 76 | ||
| AY622855.1 | - | 2 - 3 | 150 | 76 | ||
| BC014713.1 | - | 3 - 4 | 583 | 4239015 | 76 | |
| L19622.1 | - | 3 - 4 | 360 | 76 | ||
| L27424.1 | - | 3 - 4 | 630 | 76 | ||
| Z30970.1 | - | 3 - 4 | 635 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Timp3
Genomic Map