Assay Details
| Gene Symbol: Rtcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145422.4 | NP_663397.1 | 6 - 7 | 742 | 80 | |
| GenBank mRNA | AK145891.1 | - | 6 - 7 | 739 | 80 | |
| AK145916.1 | - | 6 - 7 | 739 | 80 | ||
| AK149885.1 | - | 6 - 7 | 697 | 80 | ||
| AK150376.1 | - | 6 - 7 | 701 | 80 | ||
| AK150470.1 | - | 6 - 7 | 701 | 80 | ||
| AK151042.1 | - | 6 - 7 | 735 | 80 | ||
| AK151182.1 | - | 6 - 7 | 701 | 80 | ||
| AK151932.1 | - | 6 - 7 | 701 | 80 | ||
| AK151988.1 | - | 6 - 7 | 735 | 80 | ||
| AK152144.1 | - | 6 - 7 | 701 | 80 | ||
| AK153434.1 | - | 6 - 7 | 701 | 80 | ||
| AK167643.1 | - | 6 - 7 | 728 | 80 | ||
| AK168038.1 | - | 6 - 7 | 736 | 80 | ||
| AK172175.1 | - | 6 - 7 | 736 | 80 | ||
| AK206521.1 | - | 2 - 3 | 284 | 80 | ||
| AK218642.1 | - | 2 - 3 | 284 | 80 | ||
| AY424364.1 | - | 6 - 7 | 735 | 80 | ||
| BC003288.1 | - | 6 - 7 | 744 | 3583081 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rtcb
Genomic Map