Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00017307.mm.1 |
NM_027382.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017307.mm.1 |
TC0X00002741.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00017305.mm.1 |
NM_027382.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017305.mm.1 |
TC0X00002741.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00017308.mm.1 |
NM_027382.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017308.mm.1 |
TC0X00002741.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00017309.mm.1 |
NM_027382.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017309.mm.1 |
TC0X00002741.mm.1 |
MTA TC |