Assay Details
| Gene Symbol: Hdac8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027382.3 | NP_081658.1 | 3 - 4 | 385 | 91 | |
| GenBank mRNA | AK011332.1 | - | 3 - 4 | 386 | 91 | |
| AK034511.1 | - | 3 - 4 | 351 | 91 | ||
| AK041965.1 | - | 3 - 4 | 384 | 91 | ||
| AK131998.1 | - | 3 - 4 | 598 | 91 | ||
| BC061257.1 | - | 3 - 4 | 357 | 30251620 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hdac8
Genomic Map