| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00006289.mm.1 |
NM_001177779.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00006289.mm.1 |
NM_001177779.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00006289.mm.1 |
TC0X00000963.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00006292.mm.1 |
NM_001177778.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00006292.mm.1 |
NM_001177779.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00006292.mm.1 |
NM_001177779.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00006292.mm.1 |
NM_016747.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00006292.mm.1 |
TC0X00000963.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00006290.mm.1 |
NM_001177779.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00006290.mm.1 |
NM_001177779.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00006290.mm.1 |
TC0X00000963.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00006296.mm.1 |
NM_001177778.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00006296.mm.1 |
NM_001177779.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00006296.mm.1 |
NM_001177779.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00006296.mm.1 |
NM_001177780.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00006296.mm.1 |
NM_001177780.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00006296.mm.1 |
NM_016747.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00006296.mm.1 |
TC0X00000963.mm.1 |
MTA TC |