Assay Details
| Gene Symbol: Awat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081136.1 | NP_001074605.1 | 1 - 2 | 165 | 62 | |
| GenBank mRNA | AK137763.1 | - | 1 - 2 | 162 | 62 | |
| BC147680.1 | - | 1 - 2 | 136 | 9056653 | 62 | |
| BC147683.1 | - | 1 - 2 | 136 | 9056656 | 62 | |
| BC147829.1 | - | 1 - 2 | 94 | 9087546 | 62 | |
| BC147832.1 | - | 1 - 2 | 94 | 9087549 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P117729 | BC147829.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P117729 | BC147832.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P117729 | NM_001081136.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P117729 | AK137763.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P117729 | BC147680.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412220 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 50028 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 180644 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P117729 | BC147829.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P117729 | BC147832.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P117729 | NM_001081136.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P117729 | AK137763.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P117729 | BC147680.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32560261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14910042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14847477 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14797750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560215 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14666025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14797753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14702843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560208 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560251 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14965780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14910060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15045609 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14965787 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14965785 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14910040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14797748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14515517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560245 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560209 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14698317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14797743 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14965783 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32560253 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10600991 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600990 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600992 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600993 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600989 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10600988 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00006150.mm.1 | NM_001081136.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006150.mm.1 | NM_001081136.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006150.mm.1 | TC0X00000954.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00006151.mm.1 | NM_001081136.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006151.mm.1 | NM_001081136.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006151.mm.1 | TC0X00000954.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00006152.mm.1 | NM_001081136.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006152.mm.1 | NM_001081136.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006152.mm.1 | TC0X00000954.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00006149.mm.1 | NM_001081136.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006149.mm.1 | NM_001081136.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006149.mm.1 | TC0X00000954.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 457114 | - | - |

Awat1
Genomic Map