Assay Details
| Gene Symbol: Awat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081136.1 | NP_001074605.1 | 5 - 6 | 724 | 87 | |
| GenBank mRNA | BC147680.1 | - | 5 - 6 | 695 | 9056653 | 87 |
| BC147683.1 | - | 5 - 6 | 695 | 9056656 | 87 | |
| BC147829.1 | - | 5 - 6 | 653 | 9087546 | 87 | |
| BC147832.1 | - | 5 - 6 | 653 | 9087549 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Awat1
Genomic Map