Assay Details
| Gene Symbol: Rps6ka6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025949.3 | NP_080225.2 | 11 - 12 | 1158 | 142 | |
| GenBank mRNA | AK049349.1 | - | 9 - 10 | 909 | 142 | |
| BC054113.1 | - | 9 - 10 | 831 | 30015425 | 142 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rps6ka6
Genomic Map