Assay Details
| Gene Symbol: Rps6ka6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025949.3 | NP_080225.2 | 18 - 19 | 1869 | 101 | |
| GenBank mRNA | AK012150.2 | - | 7 - 8 | 619 | 101 | |
| AK049349.1 | - | 16 - 17 | 1620 | 101 | ||
| BC054113.1 | - | 16 - 17 | 1542 | 30015425 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rps6ka6
Genomic Map