Assay Details
| Gene Symbol: Rps6ka6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025949.3 | NP_080225.2 | 19 - 20 | 1963 | 92 | |
| GenBank mRNA | AK012150.2 | - | 8 - 9 | 713 | 92 | |
| AK049349.1 | - | 17 - 18 | 1714 | 92 | ||
| BC054113.1 | - | 17 - 18 | 1636 | 30015425 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rps6ka6
Genomic Map