Assay Details
| Gene Symbol: Gk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177352.4 | NP_796326.1 | 5 - 6 | 583 | 81 | |
| GenBank mRNA | AK049275.1 | - | 5 - 6 | 584 | 81 | |
| AK076293.1 | - | 5 - 6 | 612 | 81 | ||
| AK160477.1 | - | 5 - 6 | 638 | 81 | ||
| AK162119.1 | - | 5 - 6 | 584 | 81 | ||
| AK163031.1 | - | 5 - 6 | 579 | 81 | ||
| AK167217.1 | - | 5 - 6 | 603 | 81 | ||
| BC116302.1 | - | 5 - 6 | 580 | 40095102 | 81 | |
| BC116303.1 | - | 4 - 5 | 486 | 40095106 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gk5
Genomic Map