Assay Details
| Gene Symbol: Gk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177352.4 | NP_796326.1 | 10 - 11 | 975 | 102 | |
| GenBank mRNA | AK049275.1 | - | 10 - 11 | 976 | 102 | |
| AK076293.1 | - | 10 - 11 | 1004 | 102 | ||
| AK160477.1 | - | 10 - 11 | 1030 | 102 | ||
| AK162119.1 | - | 10 - 11 | 976 | 102 | ||
| AK163031.1 | - | 10 - 11 | 971 | 102 | ||
| AK167217.1 | - | 10 - 11 | 995 | 102 | ||
| BC116302.1 | - | 10 - 11 | 972 | 40095102 | 102 | |
| BC116303.1 | - | 9 - 10 | 878 | 40095106 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gk5
Genomic Map