Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100002165.mm.1 |
NM_001159289.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002165.mm.1 |
NM_001159289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002165.mm.1 |
NM_001242934.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002165.mm.1 |
NM_001242934.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002165.mm.1 |
NM_016707.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002165.mm.1 |
NM_016707.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002165.mm.1 |
TC1100000251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100002166.mm.1 |
NM_001159289.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002166.mm.1 |
NM_001159289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002166.mm.1 |
NM_001242934.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002166.mm.1 |
NM_001242934.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002166.mm.1 |
NM_016707.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002166.mm.1 |
NM_016707.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002166.mm.1 |
TC1100000251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100002167.mm.1 |
NM_001159289.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002167.mm.1 |
NM_001159289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002167.mm.1 |
NM_001242934.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002167.mm.1 |
NM_001242934.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002167.mm.1 |
NM_016707.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002167.mm.1 |
NM_016707.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002167.mm.1 |
TC1100000251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100002164.mm.1 |
NM_001159289.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002164.mm.1 |
NM_001159289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002164.mm.1 |
NM_001242934.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002164.mm.1 |
NM_001242934.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002164.mm.1 |
NM_016707.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002164.mm.1 |
NM_016707.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002164.mm.1 |
TC1100000251.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100002170.mm.1 |
NM_001159289.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002170.mm.1 |
NM_001159289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002170.mm.1 |
NM_001159290.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002170.mm.1 |
NM_001159290.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002170.mm.1 |
NM_001242934.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002170.mm.1 |
NM_001242934.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002170.mm.1 |
NM_016707.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002170.mm.1 |
NM_016707.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100002170.mm.1 |
TC1100000251.mm.1 |
MTA TC |