Assay Details
| Gene Symbol: Atp7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109757.2 | NP_001103227.1 | 3 - 4 | 780 | 90 | |
| NM_009726.5 | NP_033856.3 | 3 - 4 | 780 | 90 | ||
| GenBank mRNA | AB007134.1 | - | 2 - 3 | 619 | 90 | |
| AK155643.1 | - | 3 - 4 | 729 | 90 | ||
| AK171563.1 | - | 3 - 4 | 696 | 90 | ||
| AK172209.1 | - | 3 - 4 | 696 | 90 | ||
| BC141395.1 | - | 3 - 4 | 658 | 9056008 | 90 | |
| U03434.1 | - | 3 - 4 | 688 | 90 | ||
| U03736.1 | - | 2 - 3 | 525 | 90 | ||
| U71091.1 | - | 3 - 4 | 681 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp7a
Genomic Map