Assay Details
| Gene Symbol: Atp7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109757.2 | NP_001103227.1 | 8 - 9 | 2094 | 135 | |
| NM_009726.5 | NP_033856.3 | 8 - 9 | 2091 | 135 | ||
| GenBank mRNA | AB007134.1 | - | 7 - 8 | 1930 | 135 | |
| BC141395.1 | - | 8 - 9 | 1972 | 9056008 | 135 | |
| U03434.1 | - | 8 - 9 | 1999 | 135 | ||
| U03736.1 | - | 7 - 8 | 1839 | 135 | ||
| U71091.1 | - | 8 - 9 | 1995 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp7a
Genomic Map