Assay Details
| Gene Symbol: Kif5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008448.3 | NP_032474.2 | 13 - 14 | 1791 | 87 | |
| GenBank mRNA | AK085221.1 | - | 5 - 6 | 617 | 87 | |
| BC090841.1 | - | 13 - 14 | 1767 | 30543821 | 87 | |
| L27153.1 | - | 13 - 14 | 1385 | 87 | ||
| U86090.1 | - | 13 - 14 | 1618 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif5b
Genomic Map