Assay Details
| Gene Symbol: Kif5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008448.3 | NP_032474.2 | 23 - 24 | 2966 | 65 | |
| GenBank mRNA | AK035154.1 | - | 1 - 2 | 402 | 65 | |
| BC090841.1 | - | 23 - 24 | 2942 | 30543821 | 65 | |
More Information
Set Membership: |
|

Kif5b
Genomic Map