Assay Details
| Gene Symbol: Abcb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029961.2 | NP_084237.1 | 22 - 23 | 2809 | 79 | |
| GenBank mRNA | AY766238.1 | - | 2 - 3 | 306 | 79 | |
| AY766239.1 | - | 22 - 23 | 2773 | 79 | ||
| JQ655148.1 | - | 9 - 10 | 2288 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcb5
Genomic Map