Assay Details
| Gene Symbol: Itgb1bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013712.2 | NP_038740.1 | 8 - 9 | 709 | 80 | |
| GenBank mRNA | AF140691.1 | - | 8 - 9 | 653 | 80 | |
| AK003906.1 | - | 8 - 9 | 710 | 80 | ||
| BC030035.1 | - | 8 - 9 | 684 | 1346742 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itgb1bp2
Genomic Map