Assay Details
| Gene Symbol: Ogt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290535.1 | NP_001277464.1 | 9 - 10 | 1199 | 98 | |
| NM_139144.4 | NP_631883.2 | 9 - 10 | 1396 | 98 | ||
| GenBank mRNA | AF363030.1 | - | 9 - 10 | 1161 | 98 | |
| BC038435.1 | - | 5 - 6 | 1455 | 98 | ||
| BC048239.1 | - | 5 - 6 | 1152 | 98 | ||
| BC057319.1 | - | 9 - 10 | 1319 | 6830845 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ogt
Genomic Map