Assay Details
| Gene Symbol: Kcnq5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160139.1 | NP_001153611.1 | 6 - 7 | 1394 | 73 | |
| NM_023872.3 | NP_076361.1 | 6 - 7 | 1394 | 73 | ||
| GenBank mRNA | AK147264.1 | - | 6 - 7 | 1397 | 73 | |
| AY679158.1 | - | 6 - 7 | 1033 | 73 | ||
| EU098138.1 | - | 1 - 2 | 43 | 73 | ||
More Information
Set Membership: |
|

Kcnq5
Genomic Map