Assay Details
Gene Symbol: Epb4.1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013813.1 | NP_038841.1 | 4 - 5 | 616 | 88 | |
GenBank mRNA | AB523362.1 | - | 3 - 4 | 504 | 88 | |
AB523363.1 | - | 3 - 4 | 504 | 88 | ||
AF152247.1 | - | 4 - 5 | 616 | 88 | ||
AF177146.1 | - | 3 - 4 | 491 | 88 | ||
AK031156.1 | - | 4 - 5 | 648 | 88 | ||
AK053532.1 | - | 4 - 5 | 617 | 88 | ||
BC079858.1 | - | 4 - 5 | 682 | 88 | ||
BC152548.1 | - | 4 - 5 | 581 | 40109749 | 88 | |
HG738870.1 | - | 3 - 4 | 504 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
