Assay Details
| Gene Symbol: L3mbtl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177278.5 | NP_796252.2 | 14 - 15 | 1782 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

L3mbtl4| Gene Symbol: L3mbtl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177278.5 | NP_796252.2 | 14 - 15 | 1782 | 94 | |
Set Membership: |
|