Assay Details
| Gene Symbol: Arhgap28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172964.4 | NP_766552.3 | 3 - 4 | 657 | 86 | |
| GenBank mRNA | AK087523.1 | - | 3 - 4 | 670 | 86 | |
| AK133720.1 | - | 3 - 4 | 499 | 86 | ||
| AK141901.1 | - | 3 - 4 | 607 | 86 | ||
| AK173147.1 | - | 2 - 3 | 284 | 86 | ||
| BC066788.1 | - | 3 - 4 | 647 | 30049271 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap28
Genomic Map