Assay Details
| Gene Symbol: Arhgap28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172964.4 | NP_766552.3 | 7 - 8 | 1053 | 112 | |
| GenBank mRNA | AK087523.1 | - | 7 - 8 | 1066 | 112 | |
| AK133720.1 | - | 7 - 8 | 895 | 112 | ||
| AK141901.1 | - | 7 - 8 | 1003 | 112 | ||
| AK162051.1 | - | 1 - 2 | 93 | 112 | ||
| AK173147.1 | - | 6 - 7 | 680 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK162051.1 transcripts.
Set Membership: |
|

Arhgap28
Genomic Map