Assay Details
Gene Symbol: Mtcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114098.1 | NP_001107570.1 | 9 - 10 | 3941 | 64 | |
NM_172963.4 | NP_766551.3 | 9 - 10 | 3939 | 64 | ||
GenBank mRNA | AK051750.1 | - | 4 - 5 | 1609 | 64 | |
AK147205.1 | - | 9 - 10 | 3941 | 64 | ||
AK147631.1 | - | 9 - 10 | 3933 | 64 | ||
AK147691.1 | - | 9 - 10 | 3933 | 64 | ||
BC032156.2 | - | 5 - 6 | 1061 | 64 | ||
BC060225.1 | - | 7 - 8 | 2187 | 30356997 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
