Assay Details
| Gene Symbol: Abcc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033336.3 | NP_001028508.2 | 23 - 24 | 3026 | 60 | |
| NM_001163675.1 | NP_001157147.1 | 23 - 24 | 3008 | 60 | ||
| NM_001163676.1 | NP_001157148.1 | 22 - 23 | 2801 | 60 | ||
| GenBank mRNA | AK032802.1 | - | 5 - 6 | 589 | 60 | |
| AK052778.1 | - | 23 - 24 | 3010 | 60 | ||
| AK157723.1 | - | 23 - 24 | 3013 | 60 | ||
| BC150822.1 | - | 23 - 24 | 2980 | 9087733 | 60 | |
| BC171974.1 | - | 22 - 23 | 2755 | 9054260 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcc4
Genomic Map