Assay Details
| Gene Symbol: Slc30a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252478.1 | NP_001239407.1 | 12 - 13 | 890 | 91 | |
| NM_144798.6 | NP_659047.2 | 12 - 13 | 875 | 91 | ||
| GenBank mRNA | AF233346.1 | - | 11 - 12 | 827 | 91 | |
| AF395840.1 | - | 11 - 12 | 817 | 91 | ||
| AK082907.1 | - | 12 - 13 | 852 | 91 | ||
| AK150342.1 | - | 12 - 13 | 851 | 91 | ||
| AK151120.1 | - | 12 - 13 | 851 | 91 | ||
| AK160903.1 | - | 12 - 13 | 879 | 91 | ||
| BC005753.1 | - | 4 - 5 | 338 | 91 | ||
| BC066162.1 | - | 12 - 13 | 842 | 30471027 | 91 | |
| BC068169.1 | - | 12 - 13 | 864 | 30542277 | 91 | |
More Information
Set Membership: |
|

Slc30a6
Genomic Map