Assay Details
| Gene Symbol: Man2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008549.2 | NP_032575.2 | 18 - 19 | 2935 | 75 | |
| GenBank mRNA | BC138371.1 | - | 18 - 19 | 2920 | 8861391 | 75 |
| BC138372.1 | - | 18 - 19 | 2920 | 8861392 | 75 | |
| X61172.1 | - | 18 - 19 | 2935 | 75 | ||
More Information
Set Membership: |
|

Man2a1
Genomic Map