Assay Details
| Gene Symbol: Tmtc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028651.2 | NP_082927.1 | 10 - 11 | 1468 | 68 | |
| GenBank mRNA | AK028333.1 | - | 10 - 11 | 1449 | 68 | |
| AK033947.1 | - | 10 - 11 | 1469 | 68 | ||
| AK036741.1 | - | 9 - 10 | 1244 | 68 | ||
| AK043293.1 | - | 10 - 11 | 1495 | 68 | ||
| AK044391.1 | - | 10 - 11 | 1424 | 68 | ||
| AK045271.1 | - | 10 - 11 | 1430 | 68 | ||
| AK049748.1 | - | 11 - 12 | 1609 | 68 | ||
| AK082526.1 | - | 9 - 10 | 1296 | 68 | ||
| BC031368.1 | - | 6 - 7 | 842 | 5029461 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmtc4
Genomic Map