Assay Details
| Gene Symbol: Tmtc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028651.2 | NP_082927.1 | 12 - 13 | 1678 | 113 | |
| GenBank mRNA | AK015064.1 | - | 2 - 3 | 256 | 113 | |
| AK017580.1 | - | 2 - 3 | 207 | 113 | ||
| AK028333.1 | - | 12 - 13 | 1659 | 113 | ||
| AK033947.1 | - | 12 - 13 | 1679 | 113 | ||
| AK036741.1 | - | 11 - 12 | 1566 | 113 | ||
| AK043293.1 | - | 12 - 13 | 1705 | 113 | ||
| AK044391.1 | - | 12 - 13 | 1634 | 113 | ||
| AK045271.1 | - | 12 - 13 | 1640 | 113 | ||
| AK049748.1 | - | 13 - 14 | 1819 | 113 | ||
| AK082526.1 | - | 11 - 12 | 1506 | 113 | ||
| BC031368.1 | - | 8 - 9 | 1052 | 5029461 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK015064.1 transcripts.
Set Membership: |
|

Tmtc4
Genomic Map