Assay Details
| Gene Symbol: Tmtc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028651.2 | NP_082927.1 | 13 - 14 | 1781 | 84 | |
| GenBank mRNA | AK015064.1 | - | 3 - 4 | 359 | 84 | |
| AK017580.1 | - | 3 - 4 | 310 | 84 | ||
| AK028333.1 | - | 13 - 14 | 1762 | 84 | ||
| AK033947.1 | - | 13 - 14 | 1782 | 84 | ||
| AK036741.1 | - | 12 - 13 | 1669 | 84 | ||
| AK043293.1 | - | 13 - 14 | 1808 | 84 | ||
| AK044391.1 | - | 13 - 14 | 1737 | 84 | ||
| AK045271.1 | - | 13 - 14 | 1743 | 84 | ||
| AK049748.1 | - | 14 - 15 | 1922 | 84 | ||
| AK082526.1 | - | 12 - 13 | 1609 | 84 | ||
| BC031368.1 | - | 9 - 10 | 1155 | 5029461 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmtc4
Genomic Map