Assay Details
| Gene Symbol: Tmtc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028651.2 | NP_082927.1 | 7 - 8 | 924 | 73 | |
| GenBank mRNA | AK028333.1 | - | 7 - 8 | 905 | 73 | |
| AK033947.1 | - | 7 - 8 | 925 | 73 | ||
| AK039093.1 | - | 7 - 8 | 869 | 73 | ||
| AK043293.1 | - | 7 - 8 | 951 | 73 | ||
| AK044391.1 | - | 7 - 8 | 880 | 73 | ||
| AK045271.1 | - | 7 - 8 | 886 | 73 | ||
| AK049748.1 | - | 8 - 9 | 1065 | 73 | ||
| AK082526.1 | - | 6 - 7 | 752 | 73 | ||
| BC031368.1 | - | 3 - 4 | 298 | 5029461 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmtc4
Genomic Map