Assay Details
| Gene Symbol: Tmtc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028651.2 | NP_082927.1 | 8 - 9 | 1082 | 82 | |
| GenBank mRNA | AK028333.1 | - | 8 - 9 | 1063 | 82 | |
| AK033947.1 | - | 8 - 9 | 1083 | 82 | ||
| AK039093.1 | - | 8 - 9 | 1027 | 82 | ||
| AK043293.1 | - | 8 - 9 | 1109 | 82 | ||
| AK044391.1 | - | 8 - 9 | 1038 | 82 | ||
| AK045271.1 | - | 8 - 9 | 1044 | 82 | ||
| AK049748.1 | - | 9 - 10 | 1223 | 82 | ||
| AK082526.1 | - | 7 - 8 | 910 | 82 | ||
| BC031368.1 | - | 4 - 5 | 456 | 5029461 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tmtc4
Genomic Map