Assay Details
| Gene Symbol: Arhgef33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145452.1 | NP_001138924.1 | 10 - 11 | 1270 | 144 | |
| GenBank mRNA | AK053944.1 | - | 6 - 7 | 904 | 144 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef33
Genomic Map