Assay Details
| Gene Symbol: Arhgef33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145452.1 | NP_001138924.1 | 11 - 12 | 1354 | 112 | |
| GenBank mRNA | AK053944.1 | - | 7 - 8 | 988 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef33
Genomic Map