Assay Details
| Gene Symbol: Ddx11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003919.1 | NP_001003919.1 | 15 - 16 | 1612 | 66 | |
| GenBank mRNA | AK049419.1 | - | 15 - 16 | 1618 | 66 | |
| AK076357.1 | - | 16 - 17 | 1642 | 66 | ||
| AK152868.1 | - | 15 - 16 | 1600 | 66 | ||
| BC049929.1 | - | 3 - 4 | 126 | 66 | ||
| BC079656.1 | - | 15 - 16 | 1612 | 6854039 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx11
Genomic Map