Assay Details
| Gene Symbol: Ddx11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003919.1 | NP_001003919.1 | 19 - 20 | 2043 | 80 | |
| GenBank mRNA | AK049419.1 | - | 19 - 20 | 2080 | 80 | |
| AK076357.1 | - | 20 - 21 | 2073 | 80 | ||
| AK152868.1 | - | 19 - 20 | 2031 | 80 | ||
| BC049929.1 | - | 7 - 8 | 557 | 80 | ||
| BC079656.1 | - | 19 - 20 | 2043 | 6854039 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx11
Genomic Map