Assay Details
| Gene Symbol: Ddx11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003919.1 | NP_001003919.1 | 23 - 24 | 2469 | 98 | |
| GenBank mRNA | AK049419.1 | - | 22 - 23 | 2619 | 98 | |
| AK152868.1 | - | 23 - 24 | 2457 | 98 | ||
| BC049929.1 | - | 11 - 12 | 983 | 98 | ||
| BC079656.1 | - | 23 - 24 | 2469 | 6854039 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx11
Genomic Map