Assay Details
| Gene Symbol: Ddx11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003919.1 | NP_001003919.1 | 24 - 25 | 2557 | 122 | |
| GenBank mRNA | AK049419.1 | - | 23 - 24 | 2707 | 123 | |
| AK076357.1 | - | 23 - 24 | 3354 | 123 | ||
| AK152868.1 | - | 24 - 25 | 2545 | 122 | ||
| BC049929.1 | - | 12 - 13 | 1071 | 122 | ||
| BC079656.1 | - | 24 - 25 | 2557 | 6854039 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx11
Genomic Map